Assay Details
Target Gene Details
Entrez Gene ID: | 100504491 |
Gene Name: | doublecortin domain containing 2b |
Gene Aliases: |
Gm12964, Gm1366, OTTMUSG00000009523 |
Location: |
Chr.4:129608331-129614477 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcdc2b | NM_001195730.1 | NP_001182659.1 |
