Assay Details
Target Gene Details
Entrez Gene ID: | 76824 |
Gene Name: | mitochondrial fission regulator 1-like |
Gene Aliases: |
2410166I05Rik, AI480750, Fam54b |
Location: |
Chr.4:134525555-134535387 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtfr1l | NM_001256112.1 | NP_001243041.1 | ||
NM_029759.4 | NP_084035.1 | |||
AK010825.1 | BAB27207.1 | |||
AK080126.1 | ||||
AK089141.1 | BAC40763.1 | |||
AK145362.1 | BAE26389.1 | |||
AK146416.1 | BAE27152.1 | |||
AK164004.1 | BAE37584.1 | |||
AK169047.1 | BAE40835.1 | |||
AK171703.1 | BAE42619.1 | |||
AK187934.1 | ||||
AK216840.1 | ||||
BC026808.1 | AAH26808.1 |
