Assay Details
Target Gene Details
Entrez Gene ID: | 329872 |
Gene Name: | Fras1 related extracellular matrix protein 1 |
Gene Aliases: |
BC037594, D430009N09, D630008K06, crf11, eye<m02Jus>, eyes2, heb, qbrick |
Location: |
Chr.4:82897920-83052548 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Frem1 | NM_001198811.1 | NP_001185740.1 | ||
NM_177863.4 | NP_808531.2 | |||
AB160986.1 | BAD89014.1 | |||
AJ616838.1 | CAE83577.1 | |||
AK052629.1 | BAC35069.1 | |||
BC138427.1 |
