Assay Details
Target Gene Details
Entrez Gene ID: | 12854 |
Gene Name: | cortistatin |
Gene Aliases: |
CST, PCST |
Location: |
Chr.4:149125191-149126741 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cort | NM_007745.3 | 1 | 72 | NP_031771.1 |
| AF013253.1 | AAB66896.1 | |||
| AK013496.1 | 1 | 74 | BAB28882.1 |
More Information
Set Membership: |
|

Genomic Map