Assay Details
Target Gene Details
Entrez Gene ID: | 56226 |
Gene Name: | espin |
Gene Aliases: |
je |
Location: |
Chr.4:152120331-152152420 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Espn | NM_207687.2 | 6 | 1087 | NP_997570.1 |
| NM_207688.2 | 2 | 149 | NP_997571.1 | |
| NM_207689.2 | 2 | 149 | NP_997572.1 | |
| NM_207690.2 | 2 | 149 | NP_997573.1 | |
| NM_207691.2 | 2 | 149 | NP_997574.2 | |
| AF239886.1 | 6 | 1087 | AAF98134.1 | |
| AF540942.1 | 2 | 149 | AAO50326.1 | |
| AF540943.1 | 2 | 149 | AAO50327.1 | |
| AF540944.1 | 2 | 149 | AAO50328.1 | |
| AF540945.1 | 2 | 149 | AAO50329.1 |
More Information
Set Membership: |
|

Genomic Map