Assay Details
Target Gene Details
Entrez Gene ID: | 57741 |
Gene Name: | nucleolar complex associated 2 homolog (S. cerevisiae) |
Gene Aliases: |
AA410003, AF155546, NIR |
Location: |
Chr.4:156235823-156247616 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Noc2l | NM_021303.2 | 2 | 95 | NP_067278.2 |
| AK076287.1 | 2 | 55 | BAC36289.1 | |
| AK078560.1 | 2 | 71 | ||
| AK133834.1 | 2 | 95 | BAE21871.1 | |
| AK157765.1 | 2 | 87 | ||
| BC020013.1 | 2 | 75 | AAH20013.1 |

Genomic Map