Assay Details
Target Gene Details
Entrez Gene ID: | 12913 |
Gene Name: | cAMP responsive element binding protein 3 |
Gene Aliases: |
AU044960, AW538053, C80076, LZIP, LZIP-1, LZIP-2, Luman |
Location: |
Chr.4:43562634-43567060 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Creb3 | NM_013497.3 | NP_038525.2 | ||
AK090077.1 | ||||
AK132785.1 | BAE21359.1 | |||
AK144208.1 | BAE25771.1 | |||
BC002094.1 | AAH02094.1 |
