Assay Details
Target Gene Details
Entrez Gene ID: | 230073 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 58 |
Gene Aliases: |
6430573D20Rik, C330021E21, RIG-I, RLR-1 |
Location: |
Chr.4:40203777-40239825 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx58 | NM_172689.3 | 16 | 2395 | NP_766277.3 |
AK049305.1 | 7 | 986 | BAC33670.1 | |
AK052595.1 | ||||
AK128929.1 | 16 | 2330 | ||
AK155930.1 | 15 | 3287 | BAE33508.1 | |
AY553221.1 | 16 | 2271 | AAS59532.1 | |
BC027369.1 | ||||
BC106095.1 | AAI06096.1 | |||
BC127163.1 | 16 | 2261 | ||
BC152540.1 | 16 | 2263 |
