Assay Details
Target Gene Details
Entrez Gene ID: | 242425 |
Gene Name: | gamma-aminobutyric acid (GABA) B receptor, 2 |
Gene Aliases: |
GABABR2, Gb2, Gm425, Gpr51 |
Location: |
Chr.4:46663898-46991714 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabbr2 | NM_001081141.1 | NP_001074610.1 | ||
| AK132744.1 |
More Information
Set Membership: |
|

Genomic Map