Assay Details
Target Gene Details
Entrez Gene ID: | 14403 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit delta |
Gene Aliases: |
AI853201 |
Location: |
Chr.4:155384979-155398129 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabrd | NM_008072.2 | NP_032098.2 | ||
| AF326546.1 | AAL37392.1 | |||
| AF326564.1 | AAL37410.1 | |||
| BC116832.1 | ||||
| BC116836.1 | ||||
| S42882.1 | AAB22967.1 |
More Information
Set Membership: |
|

Genomic Map