Assay Details
Target Gene Details
Entrez Gene ID: | 230145 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 12 |
Gene Aliases: |
9130206E10, A630062B03Rik |
Location: |
Chr.4:47091893-47123042 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt12 | NM_172693.3 | 6 | 1171 | NP_766281.1 |
| AB175680.1 | 6 | 1151 | BAD52068.1 | |
| AK033638.1 | 6 | 1172 | BAC28401.1 | |
| AK042133.1 | 6 | 1204 | BAC31179.1 | |
| AK043977.1 | 5 | 1031 | ||
| AK082014.1 | 6 | 1217 | ||
| AK156751.1 | 6 | 1201 | BAE33836.1 | |
| BC056425.1 | 6 | 1182 | AAH56425.1 |
More Information
Set Membership: |
|

Genomic Map