Assay Details
Target Gene Details
Entrez Gene ID: | 387510 |
Gene Name: | interferon kappa |
Gene Aliases: |
- |
Location: |
Chr.4:35152055-35154019 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifnk | NM_199157.2 | 2 | 738 | NP_954608.1 |
| BC099376.1 | 2 | 738 | AAH99376.1 |
Target Gene Details
Entrez Gene ID: | 214944 |
Gene Name: | MOB kinase activator 3B |
Gene Aliases: |
8430436F23Rik, A430018A01Rik, Mobkl2b |
Location: |
Chr.4:34949074-35157495 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob3b | NM_178061.5 | NP_835162.1 | ||
| AK039841.1 | BAC30466.1 | |||
| AK087285.1 | ||||
| AK155217.1 | BAE33128.1 | |||
| AK170973.1 | BAE42152.1 | |||
| BC020028.1 | AAH20028.1 |
More Information
Set Membership: |
|

Genomic Map