Assay Details
Target Gene Details
Entrez Gene ID: | 66233 |
Gene Name: | DNA methyltransferase 1-associated protein 1 |
Gene Aliases: |
1500016M21Rik, DNMAP1, DNMTAP1, Dmtap1, mKIAA1425 |
Location: |
Chr.4:117674681-117682321 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmap1 | AK005270.1 | |||
| AK012055.1 | BAB27996.3 |

Genomic Map