Assay Details
Target Gene Details
Entrez Gene ID: | 16552 |
Gene Name: | kinesin family member 12 |
Gene Aliases: |
- |
Location: |
Chr.4:63165629-63172142 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif12 | NM_010616.2 | 15 | 1561 | NP_034746.1 |
| AB755406.1 | 15 | 1574 | ||
| AK018598.1 | 15 | 1563 | BAB31300.1 | |
| BC022225.1 | 12 | 2317 | AAH22225.1 |
More Information
Set Membership: |
|

Genomic Map