Assay Details
Target Gene Details
Entrez Gene ID: | 16552 |
Gene Name: | kinesin family member 12 |
Gene Aliases: |
- |
Location: |
Chr.4:63165629-63172142 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif12 | NM_010616.2 | NP_034746.1 | ||
| AB755406.1 | ||||
| AK018598.1 | BAB31300.1 | |||
| BC022225.1 | 14 | 2774 | AAH22225.1 |
More Information
Set Membership: |
|

Genomic Map