Assay Details
Target Gene Details
Entrez Gene ID: | 26562 |
Gene Name: | neurochondrin |
Gene Aliases: |
AU042419, MMS10-AE, Ms10ae, mKIAA0607, norbin |
Location: |
Chr.4:126743750-126753566 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncdn | NM_011986.4 | 4 | 1693 | NP_036116.3 |
| AK002938.1 | 2 | 298 | BAB22468.2 | |
| AK122327.1 | 2 | 1038 | BAC65609.1 | |
| AK149932.1 | 2 | 298 | ||
| AK154889.1 | 4 | 1674 | BAE32905.1 | |
| AK170502.1 | 5 | 1408 | BAE41841.1 | |
| AK181289.1 | 1 | 57 | ||
| AK181374.1 | 1 | 60 | ||
| AK184081.1 | 1 | 57 | ||
| BC017126.1 | 4 | 1624 | AAH17126.1 |
More Information
Set Membership: |
|

Genomic Map