Assay Details
Target Gene Details
Entrez Gene ID: | 26562 |
Gene Name: | neurochondrin |
Gene Aliases: |
AU042419, MMS10-AE, Ms10ae, mKIAA0607, norbin |
Location: |
Chr.4:126743750-126753566 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncdn | NM_011986.4 | NP_036116.3 | ||
| AK002938.1 | BAB22468.2 | |||
| AK122327.1 | BAC65609.1 | |||
| AK149932.1 | ||||
| AK154889.1 | BAE32905.1 | |||
| AK170502.1 | BAE41841.1 | |||
| BC017126.1 | AAH17126.1 |
More Information
Set Membership: |
|

Genomic Map