Assay Details
Target Gene Details
Entrez Gene ID: | 69352 |
Gene Name: | N-terminal EF-hand calcium binding protein 1 |
Gene Aliases: |
1700003H21Rik, Efcbp1, STIP-1 |
Location: |
Chr.4:14952245-15149804 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Necab1 | NM_178617.4 | 10 | 1026 | NP_848732.3 |
| AK039240.1 | 10 | 842 | BAC30290.1 | |
| AK050307.1 | 11 | 923 | BAC34179.1 | |
| AK081951.1 | 10 | 918 | BAC38379.1 | |
| AY278200.2 | 10 | 1009 | AAP32077.1 | |
| BC067055.1 | 10 | 1026 | AAH67055.1 |

Genomic Map