Assay Details
Target Gene Details
Entrez Gene ID: | 69352 |
Gene Name: | N-terminal EF-hand calcium binding protein 1 |
Gene Aliases: |
1700003H21Rik, Efcbp1, STIP-1 |
Location: |
Chr.4:14952245-15149804 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Necab1 | NM_178617.4 | NP_848732.3 | ||
AK039240.1 | BAC30290.1 | |||
AK050307.1 | BAC34179.1 | |||
AK081951.1 | BAC38379.1 | |||
AY278200.2 | AAP32077.1 | |||
BC067055.1 | AAH67055.1 |
