Assay Details
Target Gene Details
Entrez Gene ID: | 230596 |
Gene Name: | PRP38 pre-mRNA processing factor 38 (yeast) domain containing A |
Gene Aliases: |
2410002M20Rik |
Location: |
Chr.4:108564867-108579336 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf38a | NM_172697.3 | NP_766285.2 | ||
| AK075925.1 | BAC36057.1 | |||
| AK169741.1 | BAE41341.1 | |||
| BC059875.1 | 1 | 337 | AAH59875.1 | |
| BC139277.1 | ||||
| BC139305.1 | ||||
| CT010186.1 | CAJ18394.1 |

Genomic Map