Assay Details
Target Gene Details
Entrez Gene ID: | 70052 |
Gene Name: | PRP4 pre-mRNA processing factor 4 homolog (yeast) |
Gene Aliases: |
1600015H11Rik, AI874830, AW047464, bN189G18.1 |
Location: |
Chr.4:62408767-62426990 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf4 | NM_027297.3 | 2 | 166 | NP_081573.1 |
| AK005464.1 | 2 | 145 | BAB24055.1 | |
| AK085174.1 | 2 | 129 | ||
| AK089838.1 | 2 | 118 | BAC40976.1 | |
| AK090114.1 | 2 | 143 | BAC41100.1 | |
| AK166912.1 | 2 | 152 | BAE39112.1 | |
| AK167216.1 | 2 | 144 | BAE39342.1 | |
| AK169532.1 | 2 | 129 | BAE41214.1 | |
| BC106089.1 | 2 | 128 | AAI06090.1 | |
| BC125529.1 | 2 | 117 | ||
| BC125531.1 | 2 | 117 |
More Information
Set Membership: |
|

Genomic Map