Assay Details
Target Gene Details
Entrez Gene ID: | 70052 |
Gene Name: | PRP4 pre-mRNA processing factor 4 homolog (yeast) |
Gene Aliases: |
1600015H11Rik, AI874830, AW047464, bN189G18.1 |
Location: |
Chr.4:62408767-62426990 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf4 | NM_027297.3 | 3 | 350 | NP_081573.1 |
| AK005464.1 | 3 | 329 | BAB24055.1 | |
| AK085174.1 | 3 | 313 | ||
| AK089838.1 | 3 | 302 | BAC40976.1 | |
| AK090114.1 | 3 | 327 | BAC41100.1 | |
| AK166912.1 | 3 | 336 | BAE39112.1 | |
| AK167216.1 | 3 | 328 | BAE39342.1 | |
| AK169532.1 | 3 | 313 | BAE41214.1 | |
| BC106089.1 | 3 | 312 | AAI06090.1 | |
| BC125529.1 | 3 | 301 | ||
| BC125531.1 | 3 | 301 |
More Information
Set Membership: |
|

Genomic Map