Assay Details
Target Gene Details
Entrez Gene ID: | 70052 |
Gene Name: | PRP4 pre-mRNA processing factor 4 homolog (yeast) |
Gene Aliases: |
1600015H11Rik, AI874830, AW047464, bN189G18.1 |
Location: |
Chr.4:62408767-62426990 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf4 | NM_027297.3 | NP_081573.1 | ||
| AK005464.1 | BAB24055.1 | |||
| AK085174.1 | ||||
| AK089838.1 | BAC40976.1 | |||
| AK090114.1 | BAC41100.1 | |||
| AK166912.1 | BAE39112.1 | |||
| AK167216.1 | BAE39342.1 | |||
| AK169532.1 | BAE41214.1 | |||
| BC106089.1 | AAI06090.1 | |||
| BC125529.1 | ||||
| BC125531.1 |
More Information
Set Membership: |
|

Genomic Map