Assay Details
Target Gene Details
Entrez Gene ID: | 230718 |
Gene Name: | 5'-nucleotidase, cytosolic IA |
Gene Aliases: |
Cn1a, Gm49 |
Location: |
Chr.4:123201210-123225333 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5c1a | NM_001085502.1 | 5 | 686 | NP_001078971.1 |
| BC147308.1 | 5 | 735 | ||
| BC147309.1 | 5 | 735 | ||
| BC147730.1 | 5 | 730 | ||
| BC147733.1 | 5 | 730 |

Genomic Map