Assay Details
Target Gene Details
Entrez Gene ID: | 18782 |
Gene Name: | phospholipase A2, group IID |
Gene Aliases: |
Splash, mGIID |
Location: |
Chr.4:138775735-138782143 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g2d | NM_011109.2 | NP_035239.1 | ||
| AF112983.1 | AAD51391.1 | |||
| AF124374.1 | AAD42773.1 | |||
| AF169407.1 | AAF42987.1 | |||
| AF169408.1 | AAF42988.1 | |||
| AK004232.1 | BAE43166.1 | |||
| AK018005.1 | BAB31033.1 | |||
| AK157392.1 | BAE34077.1 | |||
| AK158364.1 | BAE34473.1 | |||
| AK171797.1 | BAE42668.1 | |||
| AK172367.1 | BAE42967.1 | |||
| BC111806.1 | AAI11807.1 |
More Information
Set Membership: |
|

Genomic Map