Assay Details
Target Gene Details
Entrez Gene ID: | 12695 |
Gene Name: | InaD-like (Drosophila) |
Gene Aliases: |
Cipp, Patj |
Location: |
Chr.4:98395534-98719603 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inadl | NM_172696.2 | NP_766284.2 | ||
AK050545.1 | ||||
AK077268.1 | BAC36720.1 | |||
AK147866.1 | BAE28189.1 | |||
BC082787.1 | AAH82787.1 | |||
BC141404.1 |
