Assay Details
Target Gene Details
Entrez Gene ID: | 17175 |
Gene Name: | mannan-binding lectin serine peptidase 2 |
Gene Aliases: |
MASP-2, MAp19 |
Location: |
Chr.4:148594877-148615482 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Masp2 | NM_001003893.2 | 2 | 238 | NP_001003893.1 |
| NM_010767.3 | 2 | 238 | NP_034897.1 | |
| AA238260.1 | 2 | 226 | ||
| AB009459.1 | 2 | 228 | BAA34674.1 | |
| AI529228.1 | 2 | 239 | ||
| AJ250369.1 | 2 | 227 | CAB63701.1 | |
| AK050052.1 | 2 | 215 | BAC34052.1 | |
| AK149421.1 | 1 | 292 | BAE28864.1 | |
| BC013893.1 | 1 | 212 | AAH13893.1 | |
| BF233497.1 | 2 | 236 | ||
| Y19160.1 | 1 | 286 | CAB65247.1 | |
| Y19163.1 | 1 | 256 | CAB65250.1 |
More Information
Set Membership: |
|

Genomic Map