Assay Details
Target Gene Details
Entrez Gene ID: | 26561 |
Gene Name: | matrix metallopeptidase 23 |
Gene Aliases: |
CA-MMP |
Location: |
Chr.4:155650655-155654481 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mmp23 | NM_011985.2 | NP_036115.1 | ||
| AF085742.1 | AAC34886.1 | |||
| AK146510.1 | BAE27223.1 | |||
| BC107357.1 | AAI07358.1 | |||
| BC107358.1 | AAI07359.1 |
More Information
Set Membership: |
|

Genomic Map