Assay Details
Target Gene Details
Entrez Gene ID: | 12695 |
Gene Name: | InaD-like (Drosophila) |
Gene Aliases: |
Cipp, Patj |
Location: |
Chr.4:98395534-98719603 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inadl | NM_001005784.1 | 4 | 456 | NP_001005784.1 |
NM_172696.2 | 4 | 456 | NP_766284.2 | |
AK050545.1 | 4 | 428 | ||
AK147866.1 | 4 | 405 | BAE28189.1 | |
BC050846.1 | 4 | 390 | AAH50846.1 | |
BC141404.1 | 4 | 432 | ||
CK340630.1 | 4 | 456 |
