Assay Details
Target Gene Details
Entrez Gene ID: | 18491 |
Gene Name: | pregnancy-associated plasma protein A |
Gene Aliases: |
8430414N03Rik, IGFBP-4ase, PAG1, PAPP-A |
Location: |
Chr.4:65124174-65357509 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pappa | NM_021362.1 | NP_067337.1 | ||
| AF258461.1 | AAG09799.1 | |||
| AF439513.1 | AAM12687.1 | |||
| AF439514.1 | AAM12688.1 |
More Information
Set Membership: |
|

Genomic Map