Assay Details
Target Gene Details
Entrez Gene ID: | 18786 |
Gene Name: | phospholipase A2, activating protein |
Gene Aliases: |
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3 |
Location: |
Chr.4:94565139-94603247 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plaa | NM_172695.2 | 13 | 1967 | NP_766283.2 |
| AK019070.1 | 13 | 1973 | ||
| AK075934.1 | 13 | 1899 | BAC36064.1 | |
| AK085027.1 | 3 | 1414 | ||
| BC139355.1 | 13 | 1966 | ||
| BC139356.1 | 13 | 1966 | ||
| BC139773.1 | 13 | 1877 | ||
| DV646766.1 | 5 | 453 |
More Information
Set Membership: |
|

Genomic Map