Assay Details
Target Gene Details
Entrez Gene ID: | 18507 |
Gene Name: | paired box 5 |
Gene Aliases: |
BSAP, EBB-1, KLP, Pax-5 |
Location: |
Chr.4:44526269-44710481 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pax5 | NM_008782.2 | NP_032808.1 | ||
JX065130.1 | ||||
M97013.1 | AAA37325.1 |
