Assay Details
Target Gene Details
Entrez Gene ID: | 433813 |
Gene Name: | pseudouridylate synthase-like 1 |
Gene Aliases: |
2810021I11Rik, AI853843, D18346 |
Location: |
Chr.4:155888857-155891774 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pusl1 | NM_001033490.1 | NP_001028662.1 | ||
| AK151452.1 | BAE30412.1 | |||
| AK160236.1 | BAE35706.1 | |||
| AK185300.1 |
More Information
Set Membership: |
|

Genomic Map