Assay Details
Target Gene Details
Entrez Gene ID: | 22178 |
Gene Name: | tyrosinase-related protein 1 |
Gene Aliases: |
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa |
Location: |
Chr.4:80834208-80851736 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyrp1 | NM_001282014.1 | NP_001268943.1 | ||
| NM_001282015.1 | NP_001268944.1 | |||
| NM_031202.3 | NP_112479.1 | |||
| AK148332.1 | BAE28487.1 | |||
| AK148370.1 | BAE28511.1 | |||
| AK148441.1 | BAE28554.1 | |||
| BC076598.1 | AAH76598.1 | |||
| X03687.1 | CAA27323.1 |
More Information
Set Membership: |
|

Genomic Map