Assay Details
Target Gene Details
Entrez Gene ID: | 22178 |
Gene Name: | tyrosinase-related protein 1 |
Gene Aliases: |
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa |
Location: |
Chr.4:80834208-80851736 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyrp1 | NM_001282014.1 | 4 | 1116 | NP_001268943.1 |
| NM_001282015.1 | 4 | 1110 | NP_001268944.1 | |
| NM_031202.3 | 4 | 1131 | NP_112479.1 | |
| AK148332.1 | 4 | 1116 | BAE28487.1 | |
| AK148370.1 | 4 | 1022 | BAE28511.1 | |
| AK148441.1 | 4 | 1022 | BAE28554.1 | |
| AK180622.1 | ||||
| AK190391.1 | ||||
| AK190783.1 | ||||
| AK191129.1 | ||||
| AK191532.1 | ||||
| AK191540.1 | ||||
| AK193049.1 | ||||
| AK193925.1 | ||||
| AK194181.1 | ||||
| AK194480.1 | ||||
| AK194755.1 | ||||
| AK196212.1 | ||||
| AK196465.1 | ||||
| AK197240.1 | ||||
| AK197400.1 | ||||
| AK197957.1 | ||||
| AK199510.1 | ||||
| AK199962.1 | ||||
| AK200845.1 | ||||
| AK201045.1 | ||||
| AK201297.1 | ||||
| AK202385.1 | ||||
| AK203202.1 | ||||
| AK203420.1 | ||||
| AK204585.1 | ||||
| AK204597.1 | ||||
| AK204783.1 | ||||
| AK205230.1 | ||||
| AK209301.1 | ||||
| AK211654.1 | ||||
| AK214877.1 | ||||
| AK219979.1 | ||||
| BC076598.1 | 4 | 1114 | AAH76598.1 | |
| X03687.1 | 4 | 1016 | CAA27323.1 |
More Information
Set Membership: |
|

Genomic Map