Assay Details
Target Gene Details
Entrez Gene ID: | 22178 |
Gene Name: | tyrosinase-related protein 1 |
Gene Aliases: |
Oca3, TRP-1, TRP1, Tyrp, b, brown, isa |
Location: |
Chr.4:80834208-80851736 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyrp1 | NM_001282014.1 | 6 | 1483 | NP_001268943.1 |
| NM_001282015.1 | 6 | 1477 | NP_001268944.1 | |
| NM_031202.3 | 6 | 1498 | NP_112479.1 | |
| AK148332.1 | 6 | 1483 | BAE28487.1 | |
| AK148370.1 | 6 | 1389 | BAE28511.1 | |
| AK148441.1 | 6 | 1389 | BAE28554.1 | |
| AK177043.1 | ||||
| AK178803.1 | ||||
| AK180622.1 | ||||
| AK181989.1 | ||||
| AK188091.1 | ||||
| AK189039.1 | ||||
| AK189488.1 | ||||
| AK190391.1 | ||||
| AK190783.1 | ||||
| AK191129.1 | ||||
| AK191532.1 | ||||
| AK191540.1 | 3 | 398 | ||
| AK192440.1 | ||||
| AK192864.1 | ||||
| AK193049.1 | ||||
| AK193585.1 | ||||
| AK193925.1 | ||||
| AK194181.1 | ||||
| AK194427.1 | ||||
| AK194480.1 | ||||
| AK194755.1 | ||||
| AK196212.1 | ||||
| AK196465.1 | ||||
| AK197240.1 | ||||
| AK197400.1 | ||||
| AK197740.1 | ||||
| AK197957.1 | ||||
| AK198835.1 | ||||
| AK199149.1 | ||||
| AK199163.1 | ||||
| AK199296.1 | ||||
| AK199510.1 | ||||
| AK199776.1 | ||||
| AK199872.1 | ||||
| AK199962.1 | ||||
| AK200013.1 | ||||
| AK200365.1 | ||||
| AK200395.1 | ||||
| AK200845.1 | ||||
| AK201045.1 | ||||
| AK201297.1 | ||||
| AK202380.1 | ||||
| AK202385.1 | ||||
| AK203101.1 | ||||
| AK203202.1 | ||||
| AK203420.1 | ||||
| AK204301.1 | ||||
| AK204597.1 | ||||
| AK204783.1 | ||||
| AK205230.1 | ||||
| AK207921.1 | ||||
| AK209301.1 | ||||
| AK211654.1 | ||||
| AK219979.1 | ||||
| BC076598.1 | 6 | 1481 | AAH76598.1 | |
| X03687.1 | 6 | 1383 | CAA27323.1 |
More Information
Set Membership: |
|

Genomic Map