Assay Details
Target Gene Details
Entrez Gene ID: | 622404 |
Gene Name: | coiled-coil domain containing 107 |
Gene Aliases: |
- |
Location: |
Chr.4:43493340-43495921 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc107 | NM_001037913.2 | 6 | 737 | NP_001033002.1 |
AK002915.1 | 6 | 705 | BAB22454.1 | |
AK004044.1 | 6 | 723 | BAB23140.1 | |
BC027414.1 | 6 | 710 | AAH27414.1 |
