Assay Details
Target Gene Details
Entrez Gene ID: | 53378 |
Gene Name: | syndecan binding protein |
Gene Aliases: |
MDA-9, Sycl, syntenin-1 |
Location: |
Chr.4:6365680-6396122 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
More Information
Set Membership: |
|

Genomic Map