Assay Details
Target Gene Details
Entrez Gene ID: | 50780 |
Gene Name: | regulator of G-protein signaling 3 |
Gene Aliases: |
4930506N09Rik, C2PA-RGS3, C2pa, PDZ-RGS3, RGS3S |
Location: |
Chr.4:62559838-62703022 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs3 | NM_019492.3 | NP_062365.2 | ||
AJ250999.1 | CAB66146.1 |
