Assay Details
Target Gene Details
Entrez Gene ID: | 67695 |
Gene Name: | oligosaccharyltransferase 4 homolog (S. cerevisiae) |
Gene Aliases: |
2310016E02Rik |
Location: |
Chr.5:30906516-30907807 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ost4 | NM_001134692.2 | NP_001128164.1 | ||
| NM_024460.4 | NP_077780.3 | |||
| AA563474.1 | ||||
| AK002224.2 | BAE43146.1 | |||
| AK003569.1 | BAC25041.1 |
More Information
Set Membership: |
|

Genomic Map