Assay Details
Target Gene Details
Entrez Gene ID: | 19934 |
Gene Name: | ribosomal protein L22 |
Gene Aliases: |
2700038K18Rik |
Location: |
Chr.4:152324436-152334082 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl22 | NM_001277113.1 | NP_001264042.1 | ||
| NM_001277114.1 | 1 | 76 | NP_001264043.1 | |
| NM_009079.3 | 1 | 76 | NP_033105.1 | |
| NR_102291.1 | 1 | 76 | ||
| NR_102292.1 | 1 | 76 | ||
| AA755840.1 | 1 | 77 | ||
| BC096390.1 | 1 | 85 | AAH96390.1 |
More Information
Set Membership: |
|

Genomic Map