Assay Details
Target Gene Details
Entrez Gene ID: | 21812 |
Gene Name: | transforming growth factor, beta receptor I |
Gene Aliases: |
ALK5, AU017191, Alk-5, TbetaR-I, TbetaRI |
Location: |
Chr.4:47353113-47414926 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfbr1 | NM_009370.2 | NP_033396.1 | ||
| AK007125.1 | BAB24865.1 | |||
| AK015337.1 | BAB29802.1 | |||
| AK154381.1 | BAE32548.1 | |||
| BC063260.1 | AAH63260.1 | |||
| CT010393.1 | CAJ18600.1 | |||
| D25540.1 | BAA05023.1 | |||
| D28526.1 | BAA05878.1 |
More Information
Set Membership: |
|

Genomic Map