Assay Details
Target Gene Details
Entrez Gene ID: | 75062 |
Gene Name: | splicing factor 3a, subunit 3 |
Gene Aliases: |
4930512K19Rik, 60kDa |
Location: |
Chr.4:124714861-124732423 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3a3 | NM_029157.3 | NP_083433.1 | ||
| AK015776.1 | BAB29971.1 | |||
| AK030743.1 | BAC27111.1 | |||
| AK150733.1 | BAE29808.1 | |||
| AK151141.1 | BAE30148.1 | |||
| AK151382.1 | BAE30354.1 | |||
| AK161646.1 | BAE36510.1 | |||
| AK168966.1 | BAE40771.1 | |||
| AK169324.1 | BAE41078.1 | |||
| BC009141.1 | AAH09141.1 | |||
| BC092058.1 | AAH92058.1 |
More Information
Set Membership: |
|

Genomic Map