Assay Details
Target Gene Details
Entrez Gene ID: | 67615 |
Gene Name: | ubiquitin-conjugating enzyme E2R 2 |
Gene Aliases: |
1200003M11Rik, Cdc34b, Ubc3b |
Location: |
Chr.4:41136021-41193370 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2r2 | NM_026275.4 | NP_080551.1 | ||
AJ240086.1 | CAC80335.1 | |||
AK003550.1 | BAB22850.1 | |||
AK007517.1 | BAB25085.1 | |||
AK075703.1 | BAC35899.1 | |||
AK075714.1 | BAC35904.1 | |||
AK182726.1 | ||||
AK183933.1 | ||||
AK217134.1 | ||||
BC011112.1 | AAH11112.1 |
