Assay Details
Target Gene Details
Entrez Gene ID: | 66576 |
Gene Name: | ubiquinol-cytochrome c reductase hinge protein |
Gene Aliases: |
2210416J04Rik, 2310021J10Rik, 2610041P16Rik |
Location: |
Chr.4:116066965-116075070 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcrh | NM_025641.3 | NP_079917.1 | ||
| AK003266.1 | BAB22681.1 | |||
| AK003539.1 | BAB22845.1 | |||
| AK012552.1 | BAB28312.1 | |||
| AK018712.1 | BAB31360.1 | |||
| AK019085.1 | BAB31538.1 | |||
| AK028137.1 | BAC25765.1 | |||
| AK054005.1 | BAC35617.1 | |||
| AK088185.1 | BAC40195.1 | |||
| BC011388.1 | AAH11388.1 | |||
| BY138998.1 | ||||
| BY702808.1 |
More Information
Set Membership: |
|

Genomic Map