Assay Details
Target Gene Details
Entrez Gene ID: | 22608 |
Gene Name: | Y box protein 1 |
Gene Aliases: |
1700102N10Rik, C79409, EF1A, MSY1, Nsep1, YB-1, dbpB, mYB-1a |
Location: |
Chr.4:119277327-119294659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ybx1 | NM_011732.2 | NP_035862.2 | ||
AF038994.1 | AAB94768.1 | |||
AK011890.1 | ||||
AK150821.1 | BAE29883.1 | |||
AK186561.1 | ||||
AK188049.1 | ||||
BC013450.1 | AAH13450.1 | |||
BC013620.1 | AAH13620.1 | |||
BC029747.1 | AAH29747.1 | |||
BC031472.1 | AAH31472.1 | |||
BC049977.1 | AAH49977.1 | |||
BC061634.1 | AAH61634.1 | |||
BC106143.1 | AAI06144.1 | |||
BU512458.1 | ||||
M60419.1 | AAA40577.1 | |||
M62867.1 | AAA63390.1 | |||
U33196.1 | AAA75475.1 | |||
U33197.1 | AAA75476.1 | |||
X57621.1 | CAA40847.1 |
