Assay Details
Target Gene Details
Entrez Gene ID: | 68592 |
Gene Name: | SYF2 homolog, RNA splicing factor (S. cerevisiae) |
Gene Aliases: |
1110018L13Rik, Cbpin, D4Bwg1551e, Gcipip, Ntc31, mp29, p29 |
Location: |
Chr.4:134930980-134937537 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syf2 | NM_026780.3 | NP_081056.1 | ||
| AK003787.1 | BAB22995.1 | |||
| AK152794.1 | BAE31500.1 | |||
| AY033432.1 | AAK54459.1 | |||
| BC051484.1 | AAH51484.1 |

Genomic Map