Assay Details
Target Gene Details
Entrez Gene ID: | 68592 |
Gene Name: | SYF2 homolog, RNA splicing factor (S. cerevisiae) |
Gene Aliases: |
1110018L13Rik, Cbpin, D4Bwg1551e, Gcipip, Ntc31, mp29, p29 |
Location: |
Chr.4:134930980-134937537 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Syf2 | NM_026780.3 | 4 | 332 | NP_081056.1 |
AK003787.1 | 4 | 334 | BAB22995.1 | |
AK152794.1 | 4 | 321 | BAE31500.1 | |
AY033432.1 | 4 | 333 | AAK54459.1 | |
BC051484.1 | 4 | 326 | AAH51484.1 |
