Assay Details
Target Gene Details
Entrez Gene ID: | 230119 |
Gene Name: | zinc finger and BTB domain containing 5 |
Gene Aliases: |
5930421I10, 9430083K24Rik, AI646847, mKIAA0354 |
Location: |
Chr.4:44991242-45012412 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb5 | NM_001163283.1 | NP_001156755.1 | ||
| NM_001163284.1 | NP_001156756.1 | |||
| NM_173399.3 | NP_775575.2 | |||
| AY635051.1 | AAT47717.1 | |||
| BC057545.1 | AAH57545.1 |
More Information
Set Membership: |
|

Genomic Map