Assay Details
Target Gene Details
Entrez Gene ID: | 100434 |
Gene Name: | solute carrier family 44, member 1 |
Gene Aliases: |
2210409B22Rik, 4833416H08Rik, AW547365, CHTL1, CTL1, Cdw92 |
Location: |
Chr.4:53440413-53622478 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc44a1 | NM_001159633.1 | NP_001153105.1 | ||
| NM_133891.3 | NP_598652.3 | |||
| AK077951.1 | ||||
| AY249865.1 | AAP81042.1 | |||
| AY249866.1 | AAP81043.1 | |||
| BC113167.1 | ||||
| BC113169.1 | ||||
| CF532681.1 |
More Information
Set Membership: |
|

Genomic Map